Learn More
Potassium nitrate, Puratronic™, 99.995% (metals basis)
Description
Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 400°C (decomposition) |
| Melting Point | 334°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.998% (metals basis) |
| Molecular Formula | IK |
| Solubility Information | Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia. |
| InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
| SMILES | [K+].[I-] |
| Show More |
Safety and Handling
| Hazard Category | H272 |
| Precautionary Statement | P210-P220-P221-P280-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: III |
| EINECSNumber | 231-818-8 |
| RTECSNumber | TT3700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only