missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%

CAS: 7757-79-1 | KNO3 | 101.10 g/mol

Supplier:  Thermo Scientific Chemicals A145270C

 View more versions of this product

Catalog No. AAA145270C

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

Solubility
Moderately soluble in water, and insoluble in alcohol. Soluble in glycerol and ammonia.

Notes
It is an oxidizer. Stable under ordinary conditions. Incompatible with reducing agents and alcohols.
Specifications

Specifications

400°C (decomposition)
334°C
10,000 g
MFCD00011409
14,7648
Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia.
[K+].[O-][N+]([O-])=O
101.10
CHEBI:63043
99%
Hygroscopic
Potassium nitrate
7757-79-1
KNO3
UN1486
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.11
Odorless
2.109

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[K+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272
May intensify fire; oxidizer.

P210-P220-P221-P280-P370+P378q-P501c

H272

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM NITRATE

EINECSNumber : 231-818-8

RTECSNumber : TT3700000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only