missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%

Catalog No. AAA1452736
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1452736 500 g
AAA145270E 2500 g
AAA145270C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1452736 Supplier Thermo Scientific Chemicals Supplier No. A1452736

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7757-79-1 | KNO3 | 101.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

Solubility
Moderately soluble in water, and insoluble in alcohol. Soluble in glycerol and ammonia.

Notes
It is an oxidizer. Stable under ordinary conditions. Incompatible with reducing agents and alcohols.

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400°C (decomposition)
Melting Point 334°C
Quantity 500 g
UN Number UN1486
Merck Index 14,7648
Solubility Information Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia.
Formula Weight 101.11
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.109
Chemical Name or Material Potassium nitrate
Show More Show Less
Hazard Category H272
Hazard Statement GHS H Statement
H272
May intensify fire; oxidizer.
Precautionary Statement P210-P220-P221-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM NITRATE
EINECSNumber 231-818-8
RTECSNumber TT3700000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only