missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, pure, Thermo Scientific Chemicals

Catalog No. p-4380029
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446375000 500 g Glass bottle
AC446370025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446375000 Supplier Thermo Scientific Chemicals Supplier No. 446375000

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400.0°C
Melting Point 334.0°C
Quantity 500 g
Assay Percent Range 99%
Linear Formula KNO3
Merck Index 15,7769
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
IUPAC Name potassium nitrate
Formula Weight 101.1
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Potassium nitrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 231-818-8

RUO – Research Use Only