missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99+%, ACS reagent

Catalog No. AC424151000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424151000 100 g Glass bottle
AC424155000 500 g Glass bottle
AC424150025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424151000 Supplier Thermo Scientific Chemicals Supplier No. 424151000

CAS: 7757-79-1 | KNO3 | 101.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400.0°C
Melting Point 334.0°C
Color White
pH 4.5 to 8.5 (5% soln. at 25°C)
Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Linear Formula KNO3
UN Number 1486
Fieser 05,561
Merck Index 15, 7769
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
Formula Weight 101.1
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass bottle
Density 2.1
Chemical Name or Material Potassium nitrate, crystals
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
EINECSNumber 231-818-8
RTECSNumber TT3700000
TSCA TSCA

RUO – Research Use Only