missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, 1.00 Molar, Ricca Chemical
Supplier: Ricca Chemical Company R6319500500A
Specifications
| Approximately 100°C | |
| Data not available. | |
| 7732-18-5,7757-79-1,7757-79-1,7732-18-5 | |
| 90.48%,9.52% | |
| 500 mL | |
| Laboratory | |
| Colorless Liquid |
| Potassium Nitrate | |
| Colorless | |
| 90.48%,9.52% | |
| Liquid | |
| 101.1 g/mol | |
| 500 mL HDPE Bottle |
Chemical Identifiers
| 7732-18-5 | |
| 101.1 g/mol |
Safety and Handling
ShelfLife : 18 Months