missing translation for 'onlineSavingsMsg'
Learn More

Potassium molybdenum oxide, anhydrous, 99.8% (metals basis)

CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol

$90.92 - $337.18

Chemical Identifiers

CAS 13446-49-6
Molecular Formula K2MoO4
Molecular Weight (g/mol) 238.14
MDL Number MFCD00011370
InChI Key NUYGZETUIUXJCO-UHFFFAOYSA-N
Synonym potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
PubChem CID 6093816
SMILES [K+].[K+].[O-][Mo]([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2289809
View Documents
Thermo Scientific Chemicals
2289809
10 g
Each for $90.92
Only null left
 
AA2289818
View Documents
Thermo Scientific Chemicals
02289818
50 g
Each for $337.18
Only null left
 
Description

Description

Potassium molybdenum oxide used in chemical research, Chemicals, pharmaceutical research, pharmaceutical intermediates reagents, Laboratory chemicals, Manufacture of substances. And it is an important raw material and intermediates used in organic synthesis, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13446-49-6
238.14
NUYGZETUIUXJCO-UHFFFAOYSA-N
6093816
K2MoO4
MFCD00011370
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
[K+].[K+].[O-][Mo]([O-])(=O)=O

Specifications

919°C
White
10 g
K2MoO4
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
NUYGZETUIUXJCO-UHFFFAOYSA-N
dipotassium dioxomolybdenumbis(olate)
6093816
Odorless
2.3
13446-49-6
Powder
99.8% (metals basis)
MFCD00011370
Soluble in water.
[K+].[K+].[O-][Mo]([O-])(=O)=O
238.14
238.14
White powder
Potassium molybdenum oxide, anhydrous
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-599-2

RTECSNumber : QA4930000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only