missing translation for 'onlineSavingsMsg'
Learn More

Potassium molybdenum oxide, 95%

Catalog No. p-7080284
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4811222 100 g
AA4811236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4811222 Supplier Thermo Scientific Chemicals Supplier No. 04811222
Only null left

CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol

Used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-49-6
Molecular Formula K2MoO4
Molecular Weight (g/mol) 238.14
MDL Number MFCD00011370
InChI Key NUYGZETUIUXJCO-UHFFFAOYSA-N
Synonym potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
PubChem CID 6093816
IUPAC Name dipotassium;dioxido(dioxo)molybdenum
SMILES [K+].[K+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 919°C
Physical Form Powder
Quantity 100 g
Merck Index 14,7647
Solubility Information Soluble in water; Insoluble in alcohol
Formula Weight 238.14
Percent Purity 95%
Odor Odorless
Sensitivity Hygroscopic
Density 2.3 g/mL
Chemical Name or Material Potassium molybdenum oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-599-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only