missing translation for 'onlineSavingsMsg'
Learn More

Potassium Methanesulfonate 98.0+%, TCI America™

Catalog No. p-7132231
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M055025G 25 g
M0550100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M055025G Supplier TCI America Supplier No. M055025G
Only null left

Chemical Identifiers

CAS 2386-56-3
Molecular Formula CH3KO3S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00070544
InChI Key XWIJIXWOZCRYEL-UHFFFAOYSA-M
Synonym potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
PubChem CID 23666501
IUPAC Name potassium;methanesulfonate
SMILES CS(=O)(=O)[O-].[K+]

Specifications

Color White
Quantity 25 g
Formula Weight 134.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Methanesulfonate