Learn More
Potassium methanesulfonate, 99%
CAS: 2386-56-3 | CH3KO3S | 134.19 g/mol
Supplier: Thermo Scientific Chemicals 03950522
Description
Used in pharmaceutical, chemical and biological research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium methanesulfonate | |
| White | |
| CH3KO3S | |
| MFCD00070544 | |
| 3914899 | |
| potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1 | |
| XWIJIXWOZCRYEL-UHFFFAOYSA-M | |
| potassium;methanesulfonate | |
| 23666501 | |
| 99% |
| 2386-56-3 | |
| Odorless | |
| KCH3SO3 | |
| 100 g | |
| Hygroscopic | |
| Soluble in water. | |
| CS(=O)(=O)[O-].[K+] | |
| 134.19 | |
| 134.19 | |
| Powder or Lumps |
Chemical Identifiers
| 2386-56-3 | |
| 134.19 | |
| XWIJIXWOZCRYEL-UHFFFAOYSA-M | |
| 23666501 | |
| CS(=O)(=O)[O-].[K+] |
| CH3KO3S | |
| MFCD00070544 | |
| potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1 | |
| potassium;methanesulfonate |
Safety and Handling
EINECSNumber : 219-202-7
RTECSNumber : PB2675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only