missing translation for 'onlineSavingsMsg'
Learn More

Potassium methanesulfonate, 99%

Catalog No. AA3950514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3950514 25 g
AA3950522 100 g
AA3950536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3950514 Supplier Thermo Scientific Chemicals Supplier No. 03950514

CAS: 2386-56-3 | CH3KO3S | 134.19 g/mol

Used in pharmaceutical, chemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2386-56-3
Molecular Formula CH3KO3S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00070544
InChI Key XWIJIXWOZCRYEL-UHFFFAOYSA-M
Synonym potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
PubChem CID 23666501
IUPAC Name potassium;methanesulfonate
SMILES CS(=O)(=O)[O-].[K+]

Specifications

Color White
Odor Odorless
Linear Formula KCH3SO3
Quantity 25 g
Beilstein 3914899
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 134.19
Percent Purity 99%
Physical Form Powder or Lumps
Chemical Name or Material Potassium methanesulfonate
Show More Show Less
EINECSNumber 219-202-7
RTECSNumber PB2675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only