missing translation for 'onlineSavingsMsg'
Learn More

Potassium (Iodomethyl)trifluoroborate 98.0+%, TCI America™

Catalog No. P18081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18081G 1 g
P18085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18081G Supplier TCI America Supplier No. P18081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 888711-47-5
Molecular Formula CH2BF3IK
Molecular Weight (g/mol) 247.83
MDL Number MFCD11112187
InChI Key WQJVWRYIXQXISQ-UHFFFAOYSA-N
Synonym potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
PubChem CID 23674071
IUPAC Name potassium trifluoro(iodomethyl)boranuide
SMILES [K+].F[B-](F)(F)CI

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 1 g
Formula Weight 247.83
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (Iodomethyl)trifluoroborate