missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate (Powder/Certified ACS), Fisher Chemical™

Catalog No. P253100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P253100 100 g Poly Bottle
P253500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P253100 Supplier Thermo Fisher Scientific Supplier No. P253 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Melting Point 560°C
Color White
Physical Form Powder/Solid
Quantity 100 g
Assay Percent Range 99.4 to 100.4 %
Formula Weight 214
Percent Purity 99.4 to 100.4%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 3.930g/cm³
Chemical Name or Material Potassium Iodate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 5.1, : Oxidizer