missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™

Catalog No. LC196804
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC196804 4 L
LC196801 500 mL
LC196802 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC196804 Supplier LabChem Supplier No. LC196804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless
CAS Max % 96.04, 0.36, 3.5, 0.1
CAS 7758-05-6, 7681-11-0, 1310-73-2
Physical Form Liquid
Quantity 4 L
Linear Formula KIO3
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 214
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000N ±0.0005N (0.0167M)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Iodine vapor; Carbon dioxide; Carbon monoxide
Show More Show Less