missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, Solstice

Catalog No. 6001276
Change view
Click to view available options
Quantity:
250 g
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001276 250 g
6001277 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001276 Supplier Solstice Supplier No. 30314250G

Puriss. p.a., 99.7-100.4%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Quantity 250 g
Linear Formula KIO3
UN Number 1479
Formula Weight 214g/mol
Percent Purity 99.7 to 104%
Grade puriss. p.a.
Density 3.93g/mL at 25°C
Chemical Name or Material Potassium Iodate
Hazard Statement P220-P261-P305 + P351 + P338
Precautionary Statement H272-H315-H319-H335
EINECSNumber 231-831-9
RTECSNumber NN1350000