missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, ACS Reagent, Reagents

Catalog No. p-200021085
Change view
Click to view available options
Quantity:
100 mL
500 g
2.5 kg
Catalog No. Quantity
01337835 100 mL
01337834 2.5 kg
01337833 500 g
3 options

Catalog No. 01337835

Supplier: Labchem Inc C1285500100A6

Only null left

Potassium Iodate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 mL
Grade ACS
Packaging Poly Bottle
Specific Gravity 3.89
Chemical Name or Material Potassium Iodate, ACS Reagent