missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, ACS Reagent, Reagents

Catalog No. 01337835
Change view
Click to view available options
Quantity:
100 mL
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337835 100 mL
01337834 2.5 kg
01337833 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337835 Supplier Labchem Inc Supplier No. C1285500100A6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Iodate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 mL
Grade ACS
Packaging Poly Bottle
Specific Gravity 3.89
Chemical Name or Material Potassium Iodate, ACS Reagent