missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, ACS Reagent, 99.5%, Honeywell™ Fluka™
ACS Reagent, 99.5%
$223.18 - $223.18
Chemical Identifiers
| CAS | 5-6-7758 |
|---|---|
| Molecular Formula | IKO3 |
| Molecular Weight (g/mol) | 214.00 |
| MDL Number | MFCD00011406 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
| PubChem CID | 23665710 |
| IUPAC Name | potassium;iodate |
| SMILES | [O-]I(=O)=O.[K+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 5-6-7758 | |
| 214.00 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 | |
| [O-]I(=O)=O.[K+] |
| IKO3 | |
| MFCD00011406 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| potassium;iodate |
Specifications
| 560°C | |
| Colorless | |
| Solid | |
| IKO3 | |
| MFCD00011406 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| 214g/mol | |
| ACS Reagent | |
| Crystals | |
| Potassium Iodate |
| 5-6-7758 | |
| 5 to 8 (at 20°C) | |
| 100 g | |
| KIO3 | |
| UN1479 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| 99.50% | |
| Poly Bottle | |
| 3.93g/mL at 25°C; Bulk Density: ca. 2400kg/cu.m |
Safety and Handling
P220-P261-P305 + P351 + P338
H272-H315-H319-H335
DOTInformation : 5.1/PGII
EINECSNumber : 231-831-9
RTECSNumber : NN1350000