missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, ACS Grade, 99.4 to 100.4%, LabChem™

Catalog No. LC195909
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC195909 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC195909 Supplier LabChem Supplier No. LC195909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Melting Point 560°C
Color White
CAS Max % 100
Physical Form Powder
Quantity 100 g
Linear Formula KIO3
UN Number UN1479
Solubility Information Soluble in water
Formula Weight 214
Percent Purity 99.4 to 100.4%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 3930kg/m3
Decomposition Information Iodine vapor
Density 3,930kg/m3
Chemical Name or Material Potassium Iodate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Wear protective gloves, protective clothing, eye protection, face protection.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-831-9
Recommended Storage Room Temperature