missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate, 98%

Catalog No. AAA1616236
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1616236 500 g
AAA1616222 100 g
AAA161620E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1616236 Supplier Thermo Scientific Chemicals Supplier No. A1616236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium iodate, 98%, IKO3, CAS Number-7758-05-6, iodic acid, potassium salt, kaliumjodat, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, potassium iodate, iodic acid hio3 , potassium salt, caswell no. 693a, potassium triodate, potassium iodine oxide kio3, unii-i139e44nhl | CAS: 7758-05-6 | IKO3 | 214.00 g/mol

Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

Solubility
Soluble in water, potassium iodide solution. Insoluble in alcohol, liquid ammonia and nitric acid.

Notes
Incompatible with aluminum, strong oxidizing agents, sulfides, peroxides, metals and reducing agents. Avoid heat, shock and friction.

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Melting Point 560°C (decomposition)
Quantity 500 g
UN Number UN1479
Merck Index 14,7642
Solubility Information Soluble in water,potassium iodide solution. Insoluble in alcohol,liquid ammonia and nitric acid.
Formula Weight 214
Percent Purity 98%
Density 3.89
Chemical Name or Material Potassium iodate
Hazard Category H272-H302-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 231-831-9
RTECSNumber NN1350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only