missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium ICP Standard, 10,000 ppm K in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PK10KN100
Specifications
| Potassium ICP Standard | |
| 7757-79-1,7697-37-2,7732-18-5 | |
| 10,000ppm K | |
| 1 mL = 10 mg K | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium;nitrate | |
| 24434 |
| Natural LDPE Bottle | |
| 89.0,6.69,2.3 | |
| 93.0,6.96,2.4 | |
| KNO3 | |
| [N+](=O)([O-])[O-].[K+] | |
| 101.102 | |
| CHEBI:63043 |