missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, Volumetric, 8.0 M KOH (8.0 N), MilliporeSigma™ Supelco™

Catalog No. 111020056
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020056 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020056 Supplier Merck Emd Millipore Supplier No. P449450ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity 8.0 M volumetric potassium hydroxide solution for precise analytical applications.

Specifications

Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3, 7732-18-5
Linear Formula KOH
MDL Number MFCD00003553
Quantity 50 mL
Molecular Weight (g/mol) 56.11
Formula Weight 56.11
Grade Not Available
Packaging Poly Bottle