missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution AR, Macron Fine Chemicals™
macron
Supplier: Avantor 667102
Specifications
| Potassium Hydroxide Solution | |
| 45% | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| 500mL | |
| potassium;hydroxide | |
| 14797 | |
| 56.11 |
| 1310-58-3 | |
| HKO | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| [OH-].[K+] | |
| 56.105 | |
| CHEBI:32035 | |
| Poly |