missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution, 45%, for HPLC, Honeywell Fluka™
for HPLC, ∼45%
Supplier: Honeywell Chemicals 035644X25ML
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Potassium Hydroxide Solution | |
| HKO | |
| MFCD00003553 | |
| 1814 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium;hydroxide | |
| 14797 | |
| 56.11g/mol |
| 1310-58-3 | |
| KOH | |
| 45% | |
| 1.450g/mL at 20°C | |
| 4 x 25 mL | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310
H290-H302-H314