missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 45%, for HPLC, Honeywell Fluka™

Catalog No. 6001258
Change view
Click to view available options
Quantity:
25 mL
100 mL
4 x 25 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001258 25 mL
6001259 4 x 25 mL
6001257 100 mL
6001260 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001258 Supplier Honeywell Chemicals Supplier No. 0356425ML
Only null left

for HPLC, ∼45%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
Concentration 45%
UN Number 1814
Density 1.450g/mL at 20°C
Quantity 25 mL
IUPAC Name potassium;hydroxide
Formula Weight 56.11g/mol
Chemical Name or Material Potassium Hydroxide Solution
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310
Precautionary Statement H290-H302-H314