missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, 0.1M, Honeywell Fluka™
Volumetric, 0.1Â M KOH (0.1N)
Supplier: Honeywell-Fluka 3512510LVP
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
| Potassium hydroxide solution | |
| KOH | |
| 0.1M | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium;hydroxide | |
| 14797 | |
| 56.11g/mol |
| HKO | |
| MFCD00003553 | |
| NONH for all modes of transport | |
| 10 L | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 |
Chemical Identifiers
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium;hydroxide |
Safety and Handling
P305 + P351 + P338