missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 85-100.5%, pellets, Honeywell Fluka™
Puriss., meets analytical specification of BP, NF, Ph. Eur., 85-100.5%, pellets
$263.58 - $3081.53
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| SMILES | [OH-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6001263
|
Honeywell Chemicals
060051KG |
1 kg |
Each for $263.58
|
|
|||||
|
6001264
|
Honeywell Chemicals
060055KG |
5 kg |
Each for $875.47
|
|
|||||
|
60044577
|
Honeywell Chemicals
060056X1KG |
6 x 1 kg |
Case of 6 Each for $1,391.76
|
|
|||||
|
60044576
|
Honeywell Chemicals
060054X5KG |
4 x 5 kg |
Case of 4 Each for $3,081.53
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 |
Specifications
| 1310-58-3 | |
| KOH | |
| 85 to 100.5% | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium;hydroxide | |
| 14797 | |
| 56.11g/mol | |
| 1mmHg (719°C) |
| HKO | |
| MFCD00003553 | |
| 1813 | |
| 1 kg | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| puriss. | |
| Potassium Hydroxide, Meets analytical specification of Ph. Eur.BP, NF |
Safety and Handling
P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H302-H314
EINECSNumber : 215-181-3
RTECSNumber : TT2100000