Learn More
Potassium hydroxide, pure, 8N solution in water
CAS: 1310-58-3 | HKO | 56.11 g/mol
$94.67 - $189.35
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| SMILES | [OH-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC380625000
|
Thermo Scientific Chemicals
380625000 |
500 mL | Plastic bottle |
Each for $94.67
|
|
||||
|
AC380620020
|
Thermo Scientific Chemicals
380620020 |
2 L | Plastic bottle |
Each for $189.35
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 |
Specifications
| 1310-58-3,7732-18-5 | |
| Liquid | |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium;hydroxide | |
| 14797 | |
| 56.11 | |
| 7.992 to 8.008N (20°C) | |
| 1.326 | |
| Potassium hydroxide |
| Colorless | |
| 500 mL | |
| KOH | |
| 15,7761 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Pure | |
| Plastic bottle | |
| 1.3260g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 215-181-3
RUO – Research Use Only