missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Pellets, NF, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Hydroxide, Pellets, NF is used as a pH modifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 50 kg |
| Percent Purity | 85% |
| Grade | NF |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Hydroxide |