missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Pellets, NF, Spectrum™ Chemical

PO180, 1310-58-3, KOH

Supplier:  Spectrum Chemical Mfg Cor PO180500GMCS6

 View more versions of this product

Catalog No. 18605602

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Hydroxide, Pellets, NF is used as a pH modifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Potassium Hydroxide
100%
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
85%
Poly Bottle
1310-58-3
HKO
6 x 500 g
[OH-].[K+]
56.11
NF

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
[OH-].[K+]
HKO
MFCD00003553
potassium hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions