missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Pellets, FCC, 85-100.5%, Spectrum™ Chemical
P1316, 1310-58-3, KOH
$561.57 - $8048.80
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| IUPAC Name | potassium hydroxide |
| SMILES | [OH-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605595
|
Spectrum Chemical Mfg Cor
P1316500GM |
500 g | Poly Bottle |
Each for $561.57
|
|
||||
|
18605597
|
Spectrum Chemical Mfg Cor
P131625KG |
2.5 kg | Poly Bottle |
Each for $1,490.23
|
|
||||
|
18605596
|
Spectrum Chemical Mfg Cor
P1316500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,513.90
|
|
||||
|
18605598
|
Spectrum Chemical Mfg Cor
P131625KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $4,684.43
|
|
||||
|
18605599
|
Spectrum Chemical Mfg Cor
P131612KG |
12 kg | Poly Pail |
Each for $3,084.83
|
|
||||
|
18605600
|
Spectrum Chemical Mfg Cor
P131650KGBL |
50 kg | Fiber Drum |
Each for $8,048.80
|
|
||||