missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Pellets, ACS, 85%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P131525KG

 View more versions of this product

Catalog No. 18612727


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1310-58-3
HKO
2.5 kg
[OH-].[K+]
56.11
ACS
Poly Bottle
1
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
85%
To Pass Test

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
[OH-].[K+]
HKO
MFCD00003553
potassium hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.