missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Pellets, ACS, 85%, Spectrum™ Chemical

Catalog No. 18612724
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
12 kg
50 kg
100 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612724 125 g Poly Bottle
18612725 500 g Poly Bottle
18612726 1 kg Poly Bottle
18612727 2.5 kg Poly Bottle
18612728 12 kg Poly Pail
18612729 50 kg Fiber Drum
18612730 100 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18612724 Supplier Spectrum Chemical Mfg Cor Supplier No. P1315125GM

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 85%
Grade ACS
Identification To Pass Test
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.