missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide/Methanol/Hexane for Fatty Acid by Gas Chromatography, Reagents

Catalog No. 01334328
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334328 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334328 Supplier Labchem Inc Supplier No. CP1172004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Hydroxide/Methanol/Hexane for Fatty Acid by Gas Chromatography, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 110-54-3 , 1310-58-3
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Quantity 4 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.7496
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide/Methanol/Hexane for Fatty Acid by Gas Chromatography