missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, in Methanol, Certified, 1.00N ±0.01N (1.0M), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 94.5, 5.5 |
| Linear Formula | KOH |
| Concentration | 1.00N ±0.01N (1.0M) |
| UN Number | UN2924 |
| Density | 0.8g/mL |
| Solubility Information | Soluble in water |
| Quantity | 1 L |
| Formula Weight | 56.11 |
| Grade | Certified |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Toxic if swallowed, in contact with skin or if inhaled. Causes skin burns and eye damage. Causes damage to organs (central nervous system, optic nerve, liver, kidneys), (dermal, oral). |
| Precautionary Statement | GHS P Statement Keep away from heat, sparks, open flames, hot surfaces. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, ventilating, lighting equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Do no eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish. Store locked up in a cool, well ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |