missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, in Isopropanol, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

Supplier:  LabChem LC195505

 View more versions of this product

Catalog No. LC195505

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
in Isopropanol
99.4,0.6
KOH
0.1000N ±0.0005N (0.1M)
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
20 L
[OH-].[K+]
56.11
CHEBI:32035
Certified
Cubitainer™
Liquid
Colorless
1310-58-3,67-63-0
HKO
MFCD00003553
UN1993
Soluble in water
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
Passes Test
Traceable to NIST

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions