missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Certified, >45.0% (w/w), LabChem™

Supplier:  LabChem LC192501

 View more versions of this product

Catalog No. LC192501


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
1310-58-3,7732-18-5
HKO
MFCD00003553
1.44g/mL
Soluble in water
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
Certified
Poly Bottle
Traceable to NIST
Colorless
55,45
KOH
UN1814
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
500 mL
[OH-].[K+]
56.11
CHEBI:32035
>45.0% (w/w)
Passes Test
1.44g/mL
Liquid

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions