Learn More
Potassium Hydroxide, Certified, 20.0% (w/v) ±0.5% (w/v), LabChem™
$76.57 - $319.07
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| IUPAC Name | potassium hydroxide |
| SMILES | [OH-].[K+] |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |
Specifications
| Colorless | |
| 81.5,18.5 | |
| KOH | |
| UN1814 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| 500 mL | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| 20.0% ±0.5% (w/v) | |
| Passes Test | |
| 1.17g/mL | |
| Liquid |
| 1310-58-3,7732-18-5 | |
| HKO | |
| MFCD00003553 | |
| 1.17g/mL | |
| Soluble in water | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| 56.11 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST | |
| Potassium Hydroxide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature