missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Certified, 0.500N ±0.003N (0.5M), LabChem™

Supplier:  LabChem LC193202

 View more versions of this product

Catalog No. LC193202


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
1310-58-3,7732-18-5
HKO
MFCD00003553
UN1814
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
1 L
[OH-].[K+]
56.11
CHEBI:32035
Certified
Poly Bottle
Traceable to NIST
Colorless
97.25,2.75
KOH
0.500N ±0.003N (0.5M)
1.02g/mL
Soluble in water
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
Passes Test
1.02g/mL
Liquid

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions