missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Supplier: LabChem LC193004
Specifications
| Potassium Hydroxide | |
| 1310-58-3,7732-18-5 | |
| HKO | |
| MFCD00003553 | |
| 1g/mL | |
| Soluble in water | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| 56.11 | |
| Passes Test | |
| 1g/mL | |
| Liquid |
| Colorless | |
| 99.4,0.6 | |
| KOH | |
| 0.1000N ±0.0005N (0.1M) | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| 4 L | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature