missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™

Catalog No. p-7071896
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001238 500 g
6001234 2.5 kg
60045314 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001238 Supplier Honeywell Chemicals Supplier No. 221473500G
Only null left

ACS Reagent, pellets,≥85%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number 1813
Quantity 500 g
IUPAC Name potassium hydroxide
Formula Weight 56.11g/mol
Percent Purity ≥85%
Grade ACS Reagent
Vapor Pressure 1mmHg (719°C)
Chemical Name or Material Potassium Hydroxide
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H302-H314
EINECSNumber 215-181-3
RTECSNumber TT2100000