missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Linear Formula | KOH |
| UN Number | 1813 |
| Quantity | 500 g |
| IUPAC Name | potassium hydroxide |
| Formula Weight | 56.11g/mol |
| Percent Purity | ≥85% |
| Grade | ACS Reagent |
| Vapor Pressure | 1mmHg (719°C) |
| Chemical Name or Material | Potassium Hydroxide |
Safety and Handling
| Hazard Statement | P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| Precautionary Statement | H290-H302-H314 |
| EINECSNumber | 215-181-3 |
| RTECSNumber | TT2100000 |