missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ACS, 85% min, K{2}CO{3} 2.0% max

Catalog No. p-4407744
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1345118 50 g
AA1345136 500 g
AA13451A3 2 kg
AA13451A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1345118 Supplier Thermo Scientific Chemicals Supplier No. 01345118
Only null left

For Use in Soap Manufacture, in Bleaching, As An Electrolyte | CAS: 1310-58-3 | HKO | 56.11 g/mol

In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagentPotassium hydroxide acts as a precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further, it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Boiling Point 1320°C to 1324°C
Melting Point 360°C
pH 13.5
Physical Form Pellets
Quantity 50 g
UN Number UN1813
Merck Index 14,7640
Solubility Information Soluble in water,alcohol,and glycerol; Slightly soluble in ether.
IUPAC Name potassium hydroxide
Formula Weight 56.11
Percent Purity ≥85%
Grade ACS
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.044
Assay K2CO3 2.0% max
Chemical Name or Material Potassium hydroxide
Show More Show Less
Hazard Category H290-H302-H314-H335
Hazard Statement GHS H Statement
H314-H302;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLID
EINECSNumber 215-181-3
RTECSNumber TT2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only