missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 45% (w/w), Analytical Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company 61905
Specifications
| Potassium Hydroxide | |
| 1310-58-3,7732-18-5 | |
| 100.0 | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| Min. 45.0% | |
| Cubitainer | |
| Liquid |
| Colorless | |
| 98.0 | |
| HKO | |
| 45% (w/w) | |
| 20 L | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Analytical | |
| Aqueous Solution |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |