missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Potassium hydroxide, 45% w/w aq. soln

Catalog No. p-7237396
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46072AP 500 mL
AA46072K2 1 L
AA46072K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46072AP Supplier Thermo Scientific™ Supplier No. 46072AP
Only null left

CAS: 1310-58-3 | HKO | 56.11 g/mol

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number UN1814
Density 1.456
Quantity 500 mL
Formula Weight 56.11
Physical Form Liquid
ShelfLife 3 Years
EINECSNumber 215-181-3
RTECSNumber TT2100000