missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 30% (w/v) Electrolyte Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P3851LT

 View more versions of this product

Catalog No. 18612709


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Phosphate Dibasic, Trihydrate
1 L
Chemical Solution
1310-58-3,7732-18-5
30% ±0.5%
Poly Bottle

Chemical Identifiers

1310-58-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.