missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 1N solution in ethanol

Catalog No. p-3771220
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393322500 250 mL Plastic bottle
AC393320010 1 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393322500 Supplier Thermo Scientific Chemicals Supplier No. 393322500
Only null left

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Name or Material Potassium hydroxide
Color Brown-Orange to Yellow
Name Note 1N solution in ethanol
Boiling Point 78.0°C
CAS 1310-58-3, 64-17-5
Melting Point -117.0°C
Molecular Formula HKO
Linear Formula KOH
Concentration 0.950 to 1.050N
MDL Number MFCD00003553
Density 0.8600g/mL
Merck Index 15, 7761
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Solubility in water: soluble.
Quantity 250 mL
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
Flash Point 12°C
ChEBI CHEBI:32035
Formula Weight 56.11
Packaging Plastic bottle
Specific Gravity 0.86
Physical Form Liquid
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use