missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 10% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company RI61301

 View more versions of this product

Catalog No. 6130-1

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide Solution
10% (w/v)
82.0,15.92
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
9.94 to 10.06% w/v
Natural Poly Bottle
Colorless
1310-58-3,7732-18-5
85.0,16.56
MFCD00003553
4 L
[OH-].[K+]
56.11
CHEBI:32035
Laboratory
Liquid

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
CHEBI:32035
[OH-].[K+]
HKO
MFCD00003553
14797
potassium hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 months

SDS Documents

Documents

Accessories
Promotions

Promotions