missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 10% w/v aq. soln.

Catalog No. p-4824894
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
Catalog No. Quantity
AA35615K2 1 L
AA35615K7 4 L
2 options

Catalog No. AA35615K2

Supplier: Thermo Scientific Chemicals 035615K2

Only null left

CAS: 1310-58-3 | HKO | 56.11 g/mol

Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Solubility Information Miscible with water and ethanol.
Formula Weight 56.11
Appearance Colorless liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.087
Chemical Name or Material Potassium hydroxide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 215-181-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only