missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.5 N Standardized, Reagents

Catalog No. 01334322
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01334322 10 L Cubitainer
01334321 1 L Poly Bottle
01334320 20 L Cubitainer
01334319 4 L Poly Bottle
01334318 4 L Cubitainer
01334317 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01334322 Supplier Labchem Inc Supplier No. CP11930010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Hydroxide, 0.5 N Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Hydroxide, 0.5 N Standardized
pH 13.7
Color Colorless
CAS 7732-18-5, 1310-58-3
Boiling Point 100°C
Molecular Formula KOH/H2O
Synonym Caustic Potash, Potash Lye
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0236
Physical Form Liquid
Show More Show Less